1,3-Dichlor-5-nitrobenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloronitrobenzene, 1,3-Dichloro-5-nitrobenzene

OR1083-5G
APOSOR1083-5GEA 59.6 EUR
APOSOR1083-5G
1,3-Dichlor-5-nitrobenzol
1,3-Dichlor-5-nitrobenzol
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
Schmelzpunkt: 62…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007211
CAS-Nummer: 618-62-2
EINECS: 210-557-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR