N-(Brommethyl)phthalimid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(Brommethyl)-1,2-benzoldicarbonsäureimid, N-(Brommethyl)phthalsäureimid
OR10834-100G
APOSOR10834-100GEA
95.7
EUR
APOSOR10834-100G
N-(Brommethyl)phthalimid 98%
N-(Brommethyl)phthalimid
Formel:
C₉H₆BrNO₂ Molecular Weight: 240.06 g/mol Siedepunkt: 410 Schmelzpunkt: 144…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005897 CAS-Nummer: 5332-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...