2-Chloro-2-ethoxyacetic acid ethyl ester

Lieferant: Apollo Scientific
Gefahr

Synonyme: Ethyl 2-chloro-2-ethoxyacetate, Ethyl chloro(ethoxy)acetate

OR10840-1G
APOSOR10840-1GEA 26.1 EUR
APOSOR10840-1G
2-Chloro-2-ethoxyacetic acid ethyl ester
2-Chloro-2-ethoxyacetic acid ethyl ester
Formel: C₆H₁₁ClO₃
Molecular Weight: 166,601 g/mol
Siedepunkt: 81…82 °C/12mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00043953
CAS-Nummer: 34006-60-5
EINECS: 251-786-9
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR