4-Methyl-m-anissäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methoxy-4-methylbenzoesäure

OR1086-5G
APOSOR1086-5GEA 21.9 EUR
APOSOR1086-5G
4-Methyl-m-anissäure
4-Methyl-m-anissäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 152…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002566
CAS-Nummer: 7151-68-0
EINECS: 230-486-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR