3-[(Dimethylcarbamoyl)amino]benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3,3-Dimethylureido)benzoic acid, 3-(3-Carboxyphenyl)-1,1-dimethylurea

OR10863-100MG
APOSOR10863-100MGEA 62.5 EUR
APOSOR10863-100MG
3-[(Dimethylcarbamoyl)amino]benzoic acid
3-[(Dimethylcarbamoyl)amino]benzoic acid
Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411196
CAS-Nummer: 896160-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR