4-Ethoxychrysoidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR10870-10G
OR10870-1G
OR10870-5G
APOSOR10870-10GEA
0
EUR
APOSOR10870-10G
APOSOR10870-1G
APOSOR10870-5G
4-Ethoxychrysoidine hydrochloride
4-Ethoxychrysoidine hydrochloride
Formel:
C₁₄H₁₆N₄O Schmelzpunkt: 215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042023 CAS-Nummer: 2313-87-3 EINECS: 219-010-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...