3-Hydroxy-4-methyl-2(3H)-thiazolethione

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Mercapto-4-methylthiazole 3-Oxide

OR10884-1G
APOSOR10884-1GEA 168 EUR
APOSOR10884-1G
3-Hydroxy-4-methyl-2(3H)-thiazolethione
3-Hydroxy-4-methyl-2(3H)-thiazolethione
Formel: C₄H₅NOS₂
Molecular Weight: 147,21 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077793
CAS-Nummer: 49762-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR