2-Indancarboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-1H-indene-2-carboxylic acid, Indan-2-carboxylic acid

OR10885-250MG
APOSOR10885-250MGEA 21.9 EUR
APOSOR10885-250MG
2-Indancarboxylic acid 95+%
2-Indancarboxylic acid
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
Schmelzpunkt: 128…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085095
CAS-Nummer: 25177-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR