3-[(Methylsulphonyl)amino]benzoic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Methylsulphonamido)benzoic acid, N-(3-Carboxyphenyl)methanesulphonamide

OR10891-1G OR10891-5G
APOSOR10891-1GEA 108 EUR
APOSOR10891-1G APOSOR10891-5G
3-[(Methylsulphonyl)amino]benzoic acid
3-[(Methylsulphonyl)amino]benzoic acid
Formel: C₈H₉NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00596197
CAS-Nummer: 28547-13-9
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR