4-[(Methylsulphonyl)amino]benzoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N-(4-Carboxyphenyl)methanesulphonamide, 4-(Methanesulphonamido)benzoic acid, 4-(Methylsulphonamido)benzoic acid
OR10892-1G
OR10892-250MG
OR10892-5G
APOSOR10892-1GEA
49.3
EUR
APOSOR10892-1G
APOSOR10892-250MG
APOSOR10892-5G
4-[(Methylsulphonyl)amino]benzoic acid
4-[(Methylsulphonyl)amino]benzoic acid
Formel:
C₈H₉NO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025052 CAS-Nummer: 7151-76-0 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...