{3-[(Methylsulphonyl)amino]phenyl}acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[(3-Carboxymethyl)phenyl]methanesulphonamide

OR10893-500MG OR10893-50MG
APOSOR10893-500MGEA 242 EUR
APOSOR10893-500MG APOSOR10893-50MG
{3-[(Methylsulphonyl)amino]phenyl}acetic acid
{3-[(Methylsulphonyl)amino]phenyl}acetic acid
Formel: C₉H₁₁NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD03411194
CAS-Nummer: 407640-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR