Ethyl 1-benzofuran-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl benzofuran-2-carboxylate

OR10935-5G
APOSOR10935-5GEA 85.7 EUR
APOSOR10935-5G
Ethyl 1-benzofuran-2-carboxylate 97%
Ethyl-1-benzofuran-2-carboxylate
Formel: C₁₁H₁₀O₃
Molecular Weight: 190.2 g/mol
Schmelzpunkt: 29…34 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03094950
CAS-Nummer: 3199-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR