Benzyl ethyl piperidine-1,3-dicarboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Benzyl 3-ethyl piperidine-1,3-dicarboxylate
OR10941-1G
APOSOR10941-1GEA
21.9
EUR
APOSOR10941-1G
Benzyl ethyl piperidine-1,3-dicarboxylate 97%
Benzyl ethyl piperidine-1,3-dicarboxylate
Formel:
C₁₆H₂₁NO₄ Molecular Weight: 291,342 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657752 CAS-Nummer: 310454-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...