3,5-Dimethyl-4-hydroxyphenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-Dimethy-4-hydroxyphenylboronic acid pinacol ester

OR10985-1G
APOSOR10985-1GEA 31.9 EUR
APOSOR10985-1G
3,5-Dimethyl-4-hydroxyphenylboronsäurepinakolester
3,5-Dimethyl-4-hydroxyphenylboronsäurepinakolester
Formel: C₁₄H₂₁BO₃
Molecular Weight: 248,13 g/mol
Schmelzpunkt: 100…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093724
CAS-Nummer: 269410-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR