2-(Ethoxycarbonyl)phenylboronsäurepinakolester 95%

Lieferant: Apollo Scientific

Synonyme: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR10987-25G OR10987-5G
APOSOR10987-25GEA 258 EUR
APOSOR10987-25G APOSOR10987-5G
2-(Ethoxycarbonyl)phenylboronsäurepinakolester 95%
2-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Schmelzpunkt: 59…62 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03453660
CAS-Nummer: 269409-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR