4-Iod-1H-pyrazol 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iodopyrazole

OR10990-100G OR10990-25G OR10990-500G
APOSOR10990-100GEA 39.3 EUR
APOSOR10990-100G APOSOR10990-25G APOSOR10990-500G
4-Iod-1H-pyrazol 99%
4-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.97 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005244
CAS-Nummer: 3469-69-0
EINECS: 222-434-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR