4-Hydroxy-3-methoxyphenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

OR10992-250MG
APOSOR10992-250MGEA 21.9 EUR
APOSOR10992-250MG
4-Hydroxy-3-methoxyphenylboronsäurepinakolester 98%
4-Hydroxy-3-methoxyphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₄
Molecular Weight: 250,099 g/mol
Schmelzpunkt: 105…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093723
CAS-Nummer: 269410-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR