4'-Methylbiphenyl-4-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

OR10996-1G
APOSOR10996-1GEA 21.9 EUR
APOSOR10996-1G
4'-Methylbiphenyl-4-carbonsäure 98%
4'-Methylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00448778
CAS-Nummer: 720-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR