3-Amino-5-nitrobenz[d]isothiazole
Lieferant: Apollo Scientific
OR110001-1G
APOSOR110001-1GEA
87
EUR
APOSOR110001-1G
3-Amino-5-nitrobenz[d]isothiazole
3-Amino-5-nitrobenz[d]isothiazole
Formel:
C₇H₅N₃O₂S Molecular Weight: 195,20 g/mol Schmelzpunkt: 250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270828 CAS-Nummer: 84387-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...