3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Lieferant: Apollo Scientific
|
Achtung
|
OR110002-1G
APOSOR110002-1GEA
125
EUR
APOSOR110002-1G
3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Formel:
C₁₅H₂₁NO₂ Schmelzpunkt: 172…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00276840 CAS-Nummer: 71556-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...