3-(Piperidin-1-yl)pyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Cyanopyridin-3-yl)piperidine, 3-(Piperidin-1-yl)picolinonitrile, 2-Cyano-3-(piperidin-1-yl)pyridine

OR110004-1G
APOSOR110004-1GEA 248 EUR
APOSOR110004-1G
3-(Piperidin-1-yl)pyridine-2-carbonitrile
3-(Piperidin-1-yl)pyridine-2-carbonitrile
Formel: C₁₁H₁₃N₃
Molecular Weight: 187,241 g/mol
Schmelzpunkt: 65…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690241
CAS-Nummer: 780802-33-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR