[2-(Morpholin-4-ylmethyl)phenyl]methanol

Lieferant: Apollo Scientific

OR110005-1G
APOSOR110005-1GEA 395 EUR
APOSOR110005-1G
[2-(Morpholin-4-ylmethyl)phenyl]methanol
[2-(Morpholin-4-ylmethyl)phenyl]methanol
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03211251
CAS-Nummer: 91271-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR