[3-(Pyridin-2-yloxy)phenyl]methanol

Lieferant: Apollo Scientific
Achtung

OR110006-1G
APOSOR110006-1GEA 183 EUR
APOSOR110006-1G
[3-(Pyridin-2-yloxy)phenyl]methanol
[3-(Pyridin-2-yloxy)phenyl]methanol
Formel: C₁₂H₁₁NO₂
Schmelzpunkt: 56…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025855
CAS-Nummer: 869901-22-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR