N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine

Lieferant: Apollo Scientific

OR110008-1G
APOSOR110008-1GEA 395 EUR
APOSOR110008-1G
N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine
N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine
Formel: C₁₂H₁₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09025859
CAS-Nummer: 879896-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR