{2-[2-(Dimethylamino)ethoxy]phenyl}methanol

Lieferant: Apollo Scientific
Gefahr

OR110009-1G
APOSOR110009-1GEA 232 EUR
APOSOR110009-1G
{2-[2-(Dimethylamino)ethoxy]phenyl}methanol
{2-[2-(Dimethylamino)ethoxy]phenyl}methanol
Formel: C₁₁H₁₇NO₂
Molecular Weight: 195,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08690289
CAS-Nummer: 59115-26-3
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR