N-Methyl-N-[2-(tetrahydro-2H-pyran-4-yloxy)benzyl]amine

Lieferant: Apollo Scientific

OR110010-1G
APOSOR110010-1GEA 395 EUR
APOSOR110010-1G
N-Methyl-N-[2-(tetrahydro-2H-pyran-4-yloxy)benzyl]amine
N-Methyl-N-[2-(tetrahydro-2H-pyran-4-yloxy)benzyl]amine
Formel: C₁₃H₁₉NO₂
Molecular Weight: 221,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09064962
CAS-Nummer: 906352-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR