(5-Morpholin-4-ylthien-2-yl)methanol

Lieferant: Apollo Scientific

OR110011-1G
APOSOR110011-1GEA 198 EUR
APOSOR110011-1G
(5-Morpholin-4-ylthien-2-yl)methanol
(5-Morpholin-4-ylthien-2-yl)methanol
Formel: C₉H₁₃NO₂S
Molecular Weight: 199.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08442796
CAS-Nummer: 910036-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR