[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol

Lieferant: Apollo Scientific
Gefahr

OR110012-1G
APOSOR110012-1GEA 173 EUR
APOSOR110012-1G
[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol
[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol
Formel: C₁₂H₁₇NO
Molecular Weight: 191,269 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08690298
CAS-Nummer: 91271-60-2
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR