3-(1H-Pyrazol-1-ylmethyl)benzaldehyd

Lieferant: Apollo Scientific

OR110014-1G
APOSOR110014-1GEA 0 EUR
APOSOR110014-1G
3-(1H-Pyrazol-1-ylmethyl)benzaldehyd
3-(1H-Pyrazol-1-ylmethyl)benzaldehyd
Formel: C₁₁H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08059836
CAS-Nummer: 78425-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR