[3-(1H-Pyrazol-1-ylmethyl)phenyl]methanol

Lieferant: Apollo Scientific

OR110015-1G
APOSOR110015-1GEA 248 EUR
APOSOR110015-1G
[3-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
[3-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08572163
CAS-Nummer: 78425-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR