3-(Phenoxymethyl)aniline
Lieferant: Apollo Scientific
|
Gefahr
|
OR110016-1G
APOSOR110016-1GEA
395
EUR
APOSOR110016-1G
3-(Phenoxymethyl)aniline
3-(Phenoxymethyl)aniline
Formel:
C₁₃H₁₃NO Schmelzpunkt: 42…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804492 CAS-Nummer: 93189-16-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...