N-{[1-(2-Furylmethyl)piperidin-4-yl]methyl}-N-methylamine

Lieferant: Apollo Scientific

OR110018-1G
APOSOR110018-1GEA 0 EUR
APOSOR110018-1G
N-{[1-(2-Furylmethyl)piperidin-4-yl]methyl}-N-methylamine
N-{[1-(2-Furylmethyl)piperidin-4-yl]methyl}-N-methylamine
Formel: C₁₂H₂₀N₂O
Molecular Weight: 208,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09966159
CAS-Nummer: 934570-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR