4-Nitro-5-piperazin-1-ylthiophene-2-sulphonamide

Lieferant: Apollo Scientific

OR110022-1G
APOSOR110022-1GEA 0 EUR
APOSOR110022-1G
4-Nitro-5-piperazin-1-ylthiophene-2-sulphonamide
4-Nitro-5-piperazin-1-ylthiophene-2-sulphonamide
Formel: C₈H₁₂N₄O₄S₂
Molecular Weight: 292,335 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00728810
CAS-Nummer: 845266-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR