(4-Methyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-Methyl-1-(piperidin-3-ylcarbonyl)piperidine hydrochloride

OR110023-1G
APOSOR110023-1GEA 0 EUR
APOSOR110023-1G
(4-Methyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Formel: C₁₂H₂₃ClN₂O
Molecular Weight: 246,78 g/mol
Schmelzpunkt: 177…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06164734
CAS-Nummer: 845885-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR