(4-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-Methyl-1-(piperidin-4-ylcarbonyl)piperidine hydrochloride

OR110024-1G
APOSOR110024-1GEA 0 EUR
APOSOR110024-1G
(4-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Formel: C₁₂H₂₃ClN₂O
Molecular Weight: 246,78 g/mol
Schmelzpunkt: 279…281 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06407989
CAS-Nummer: 690632-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR