1-Piperidinyl(2-piperidinyl)methanone hydrochloride

Lieferant: Apollo Scientific

Synonyme: 1-(Piperidin-2-ylcarbonyl)piperidine hydrochloride

OR110025-1G
APOSOR110025-1GEA 0 EUR
APOSOR110025-1G
1-Piperidinyl(2-piperidinyl)methanone hydrochloride
1-Piperidinyl(2-piperidinyl)methanone hydrochloride
Formel: C₁₁H₂₁ClN₂O
Molecular Weight: 232,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06407996
CAS-Nummer: 130497-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR