(4-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
Lieferant: Apollo Scientific
OR110026-1G
APOSOR110026-1GEA
0
EUR
APOSOR110026-1G
(4-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
Formel:
C₁₂H₂₃ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06407988 CAS-Nummer: 690634-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...