4-Cyclohexylpiperazine-1-carboximidamide hydroiodide

Lieferant: Apollo Scientific
Achtung

OR110027-1G
APOSOR110027-1GEA 79.9 EUR
APOSOR110027-1G
4-Cyclohexylpiperazine-1-carboximidamide hydroiodide
4-Cyclohexylpiperazine-1-carboximidamide hydroiodide
Formel: C₁₁H₂₂N₄·HI
Molecular Weight: 338,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368564
CAS-Nummer: 849776-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR