2,5-Dibromo-4-pyridinylboronic acid

Lieferant: Apollo Scientific

Synonyme: (2,5-Dibromopyridin-4-yl)boronic acid

OR110029-1G
APOSOR110029-1GEA 0 EUR
APOSOR110029-1G
2,5-Dibromo-4-pyridinylboronic acid
2,5-Dibromo-4-pyridinylboronic acid
Formel: C₅H₄BBr₂NO₂
Molecular Weight: 280,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109338
CAS-Nummer: 1031843-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR