Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-1-phenylpyrazole-4-carboxylic acid Ethyl Ester, 5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester, 5-Amino-1-phenylpyrazole-4-carboxylic acid ethyl ester, 5-Amino-4-carbethoxy-1-phenylpyrazole, Ethyl 5-Amino-1-phenylpyrazole-4-carboxylate, Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate

OR110031-25G OR110031-5G
APOSOR110031-25GEA 58 EUR
APOSOR110031-25G APOSOR110031-5G
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat 98%
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Formel: C₇H₁₁N₃O₂
Schmelzpunkt: 98…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020731
CAS-Nummer: 16078-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR