Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-1-phenylpyrazole-4-carboxylic acid Ethyl Ester, 5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester, 5-Amino-1-phenylpyrazole-4-carboxylic acid ethyl ester, 5-Amino-4-carbethoxy-1-phenylpyrazole, Ethyl 5-Amino-1-phenylpyrazole-4-carboxylate, Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate
OR110031-25G
OR110031-5G
APOSOR110031-25GEA
58
EUR
APOSOR110031-25G
APOSOR110031-5G
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat 98%
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Formel:
C₇H₁₁N₃O₂ Schmelzpunkt: 98…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020731 CAS-Nummer: 16078-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...