Methyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110032-1G
APOSOR110032-1GEA 69.6 EUR
APOSOR110032-1G
Methyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Methyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Formel: C₁₃H₉ClO₂S₂
Molecular Weight: 296,792 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00106778
CAS-Nummer: 255378-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR