N-(4-Ethoxycarbonylphenyl)maleamidsäure

Lieferant: Apollo Scientific

Synonyme: 4-{[4-(Ethoxycarbonyl)phenyl]amino}-4-oxobut-2-enoic acid

OR110035-5G
APOSOR110035-5GEA 36.4 EUR
APOSOR110035-5G
N-(4-Ethoxycarbonylphenyl)maleamidsäure
N-(4-Ethoxycarbonylphenyl)maleamidsäure
Formel: C₁₃H₁₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00063173
CAS-Nummer: 200126-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR