Methyl-3-brom-1H-indazol-6-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-bromo-1H-indazole-6-carboxylate

OR110044-1G
APOSOR110044-1GEA 65.4 EUR
APOSOR110044-1G
Methyl-3-brom-1H-indazol-6-carboxylat
Methyl-3-brom-1H-indazol-6-carboxylat
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,0700 g/mol
Schmelzpunkt: 172…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11499017
CAS-Nummer: 192945-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR