Ethyl 2-amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyran-3-carboxylate

OR110045-1G
APOSOR110045-1GEA 69.6 EUR
APOSOR110045-1G
Ethyl 2-amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxylate
Ethyl 2-amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxylate
Formel: C₁₀H₁₃NO₃S
Molecular Weight: 227,28 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD05664038
CAS-Nummer: 117642-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR