Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate

Lieferant: Apollo Scientific

OR110046-1G
APOSOR110046-1GEA 58 EUR
APOSOR110046-1G
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate
Formel: C₉H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02186164
CAS-Nummer: 64951-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR