1-(3-Methoxybenzyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR110047-1G
APOSOR110047-1GEA 0 EUR
APOSOR110047-1G
1-(3-Methoxybenzyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Methoxybenzyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₃H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08164335
CAS-Nummer: 96449-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR