1-(4-Bromo-3-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
Lieferant: Apollo Scientific
OR110050-1G
APOSOR110050-1GEA
186
EUR
APOSOR110050-1G
1-(4-Bromo-3-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Bromo-3-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel:
C₁₂H₁₂BrNO₃ Molecular Weight: 298,133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02647934 CAS-Nummer: 361463-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...