N-(2,4-Dichloro-6-methylphenyl)thiourea

Lieferant: Apollo Scientific

OR110052-1G
APOSOR110052-1GEA 186 EUR
APOSOR110052-1G
N-(2,4-Dichloro-6-methylphenyl)thiourea
N-(2,4-Dichloro-6-methylphenyl)thiourea
Formel: C₈H₈Cl₂N₂S
Molecular Weight: 235,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24850049
CAS-Nummer: 10524-81-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR