Methyl-6-(1H-pyrrol-1-yl)nicotinate

Lieferant: Apollo Scientific

OR110054-1G
APOSOR110054-1GEA 0 EUR
APOSOR110054-1G
Methyl-6-(1H-pyrrol-1-yl)nicotinate
Methyl-6-(1H-pyrrol-1-yl)nicotinate
Formel: C₁₁H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04973730
CAS-Nummer: 852180-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR